Ammonium metavanadate ≥99.5%, Puriss. p.a. Reag. Ph. Eur., Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ammonium monovanadate, Ammonium vanadium trioxide, Ammonium vanadium oxide

31153-25KG
HONC31153-25KGEA 14550 GBP
HONC31153-25KG
Ammonium metavanadate ≥99.5%, Puriss. p.a. Reag. Ph. Eur., Fluka™
Ammonium metavanadate
Formula: NH₄VO₃
Melting Pt: 200 °C
Density: 2.3 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00011430
CAS Number: 7803-55-6
EINECS: 232-261-3
UN: 2859
ADR: 6.1,II
Merck Index: 13,00572

Order Now

Specification Test Results

Assay Min. 99.5 %
Cadmium (Cd) Max. 0.001 %
Cobalt (Co) Max. 0.002 %
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.001 %
Nickel (Ni) Max. 0.002 %
Lead (Pb) Max. 0.002 %
Zinc (Zn) Max. 0.001 %
Chloride (Cl) Max. 0.002 %
Phosphate (PO₄) Max. 0.005 %
Sulfate (SO₄) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR