Hydrogen peroxide 34.5-36.5%, meets analytical specification of Ph. Nord.

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

18304H-5L 18304H-1L 18304H-6X1L
HONC18304H-5LEA 224 GBP
HONC18304H-5L HONC18304H-1L HONC18304H-6X1L
Hydrogen peroxide 34.5-36.5%, meets analytical specification of Ph. Nord.
Hydrogen peroxide
Formula: H₂O₂
Melting Pt: –33 °C
Density: 1.110 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1,II

Order Now

Specification Test Results

Assay 34.5 - 36.5 %
Non-volatile matter Max. 0.05 %
Residue on ignition Max. 0.03 %
Free acid (as H₂SO₄) Max. 0.02 %
Heavy metals (as Pb) Max. 0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR