Pipette controller, pipetus®-standard

Supplier: HIRSCHMANN
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

9903000 9904000 9905000 9906000 9904010 9903010
612-2602EA 320 GBP
612-2602 612-2603 HIRS9905000 HIRS9906000 HIRS9904010 HIRS9903010
Pipette controller, pipetus®-standard
Pipettes Pipette Controllers
Pipette controller for pipettes from 0,1 to 100 ml.

  • Optimum safety and excellent convenience of use
  • Pipettor is lighter and easy to use because the pump is separated from the handle
  • Autoclavable, detachable handle for the pipette holder is equipped with two buttons for filling and emptying

This electric pipettor is equipped with a 0,2 µm integrated water-repellent filter (for better durability) and a safety valve to protect the equipment from the introduction of hazardous (contaminated or radioactive) liquids.

Ordering information: The completeMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR