Tubing for rotarus® peristaltic pumps, Tygon® LFL
Supplier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
95223011 95223013 95223015 95223016 95223017
HIRS95223011EA 543 GBP
HIRS95223011 HIRS95223013 HIRS95223015 HIRS95223016 HIRS95223017
Tubing for rotarus® peristaltic pumps, Tygon® LFL
Tubing Transfer Tubing
Long flex life pump tube for general applications in the lab and industry and a long service life with maximum bending fatigue strength of all transparent Tygon® tubes, extremely low particle detachment, broad chemical resistance, long service life.
  • Temperature resistance from –54 to 74 °C
  • Hardness (Shore durometer A): 56