Tubing for rotarus® peristaltic pumps, Tygon® SPT-3350
Supplier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
95217012 95217722 95217037 95217725 95217038 95217717 95217710 95217525 95217522 95217517 95217510 95217036 95217035 95217019 95217018 95217017 95217016 95217015 95217013 95217011
HIRS95217012EA 381 GBP
HIRS95217012 HIRS95217722 HIRS95217037 HIRS95217725 HIRS95217038 HIRS95217717 HIRS95217710 HIRS95217525 HIRS95217522 HIRS95217517 HIRS95217510 HIRS95217036 HIRS95217035 HIRS95217019 HIRS95217018 HIRS95217017 HIRS95217016 HIRS95217015 HIRS95217013 HIRS95217011
Tubing for rotarus® peristaltic pumps, Tygon® SPT-3350
Tubing Transfer Tubing
Tese sanitary silicon tubes are platinum cured silicon tube, ultra smooth inner wall, non-toxic, non-haemolytic, non-pyrogenic.
  • Temperature resistance from −80 to 200 °C
  • Hardness (Shore durometer A): 50