Tubing for rotarus® peristaltic pumps, Tygon® Chemical
Supplier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
95215013 95215018 95215016 95215015
HIRS95215013EA 562 GBP
HIRS95215013 HIRS95215018 HIRS95215016 HIRS95215015
Tubing for rotarus® peristaltic pumps, Tygon® Chemical
Tubing Transfer Tubing
These tubes are used in chemical application for its very high chemical resistance to acids, bases, salts, ketones, alcohols.
  • Temperature resistance from –60 to 74 °C
  • Hardness (Shore durometer A): 61