Micro burettes, bang pattern, lateral stopcock, Schellbach
Supplier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3310140 3310152 3310160 3310340 3310352 3310360
HIRS3310140EA 344 GBP
HIRS3310140 HIRS3310152 HIRS3310160 HIRS3310340 HIRS3310352 HIRS3310360
Micro burettes, bang pattern, lateral stopcock, Schellbach
Burettes
Micro burette, class AS, DURAN®, blue graduation, certified conformity, with dated batch identification.
  • Lateral stopcock with standard ground glass key or PTFE spindle, with socket, Bang
  • Schellbach stripes for enhanced legibility
  • Main point ring graduation
Certifications: Calibrated for EX, DIN EN ISO 385.