Mixing measuring cylinders, tall form, class A, borosilicate glass
Supplier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2340293 2340294 2340260 2340270 2340275 2340280 2340286 2340290
HIRS2340293EA 227 GBP
HIRS2340293 HIRS2340294 HIRS2340260 HIRS2340270 HIRS2340275 HIRS2340280 HIRS2340286 HIRS2340290
Mixing measuring cylinders, tall form, class A, borosilicate glass
Cylinders Measuring Cylinders
DURAN®, borosilicate glass, with hollow glass stopper, class A, conformity certified.
  • Blue Graduation, main point graduation
  • Hexagonal glass base, NS ground socket
  • Conformity certified, with dated batch identification
Certifications: DIN EN ISO 4788