Antibiotic Sensitivity Discs
Supplier: HIMEDIA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SD212-5X50DS SD094-5X50DS SD725-5X50DS SD211-5X50DS SD041-5X50DS SD048-5X50DS SD110-5X50DS SD704-5X50DS SD283-5X50DS SD179-5X50DS SD115-5X50DS SD108-5X50DS SD012-5X50DS SD010-5X50DS SD753-5X50DS SD020-5X50DS SD024-5X50DS SD217-5X50DS SD224-5X50DS SD232-5X50DS SD192-5X50DS SD273-5X50DS SD002-5X50DS SD016-5X50DS SD025-5X50DS SD040-5X50DS SD047-5X50DS SD057-5X50DS SD060-5X50DS SD081-5X50DS SD087-5X50DS SD154-5X50DS SD204-5X50DS SD216-5X50DS
HIMESD212-5X50DSEA 21 GBP
HIMESD212-5X50DS HIMESD094-5X50DS HIMESD725-5X50DS HIMESD211-5X50DS HIMESD041-5X50DS HIMESD048-5X50DS HIMESD110-5X50DS HIMESD704-5X50DS HIMESD283-5X50DS HIMESD179-5X50DS HIMESD115-5X50DS HIMESD108-5X50DS HIMESD012-5X50DS HIMESD010-5X50DS HIMESD753-5X50DS HIMESD020-5X50DS HIMESD024-5X50DS HIMESD217-5X50DS HIMESD224-5X50DS HIMESD232-5X50DS HIMESD192-5X50DS HIMESD273-5X50DS HIMESD002-5X50DS HIMESD016-5X50DS HIMESD025-5X50DS HIMESD040-5X50DS HIMESD047-5X50DS HIMESD057-5X50DS HIMESD060-5X50DS HIMESD081-5X50DS HIMESD087-5X50DS HIMESD154-5X50DS HIMESD204-5X50DS HIMESD216-5X50DS
Antibiotic Sensitivity Discs
Microbiology Test Systems Antibiotic Tests
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.