Avantor® Alltima, HPLC Columns, Cyano
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HICH81189EA 996 GBP
HICH81189 HICH88180 HICH88182 HICH88189 HICH88191 96084/N AL-5CN-150DE
Avantor® Alltima, HPLC Columns, Cyano
Chromatography Columns
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L10
General purpose cyano suitable for normal or reversed phase use. Suitable for rugged normal phase applications.
Order Now
SPECIFICATIONS
Environmentally Preferable
Endcapped Yes
Surface area 340 m²/g
USP code L10