Avantor® Hichrom HI-8 HT, GC Columns
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HI88-32-010-10G2EA 741 GBP
HI88-32-010-10G2 HI88-25-010-15 HI88-25-010-30 HI88-25-015-15 HI88-25-015-30 HI88-25-025-15 HI88-25-025-30 HI88-32-010-10 HI88-32-010-15 HI88-32-010-30 HI88-32-015-15 HI88-32-015-30 HI88-32-025-15 HI88-32-025-30 HI88-53-010-10 HI88-53-010-15 HI88-53-010-30 HI88-53-015-15 HI88-53-015-30 HI88-53-017-30 HI88-53-025-15 HI88-53-025-30 HI88-10-010-10 HI88-10-010-5
Avantor® Hichrom HI-8 HT, GC Columns
Chromatography Columns
High Temperature general purpose column ideal for PCBs compounds, Pesticides, environmental analysis.
Max temperature limits may change depending on the film thickness.
Order Now
SPECIFICATIONS
Phase name HI-8 HT
Environmentally Preferable
Phase composition Proprietary Phase - high temperature
Polarity Low-to-mid polarity stationary phase
Crossbond Yes
Temperature limit 400 °C