Avantor® Hichrom HI-ALC 2, GC Columns
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HI37-32-060-30EA 982 GBP
HI37-32-060-30 HI37-32-100-30 HI37-53-100-30
Avantor® Hichrom HI-ALC 2, GC Columns
Chromatography Columns
Application specific column for blood alcohols testing.
Max temperature limits may change depending on the film thickness.