Avantor® Hichrom HI-17 MS, GC Columns
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HI22-25-010-30G5EA 757 GBP
HI22-25-010-30G5 HI22-25-025-15G5 HI22-25-025-30G10 HI22-25-025-30G5 HI22-25-025-60G10 HI22-25-025-60G5 HI22-25-050-15G10 HI22-25-050-30G10 HI22-25-050-30G5 HI22-32-025-30G5 HI22-32-100-30G5 HI22-53-100-30G5 HI22-18-018-10 HI22-18-018-20 HI22-18-018-40 HI22-18-018-50 HI22-18-018-60 HI22-18-020-10 HI22-18-020-20 HI22-18-020-40 HI22-18-020-50 HI22-18-020-60 HI22-18-036-20 HI22-18-036-40 HI22-18-036-60 HI22-18-040-10 HI22-18-040-20 HI22-18-040-40 HI22-18-040-50 HI22-18-040-60 HI22-20-010-10 HI22-20-010-20 HI22-20-010-40 HI22-20-010-50 HI22-20-010-60 HI22-20-020-10 HI22-20-020-20 HI22-20-020-40 HI22-20-020-50 HI22-20-020-60 HI22-20-033-12 HI22-20-033-25 HI22-20-033-30 HI22-20-033-50 HI22-20-033-60 HI22-20-040-10 HI22-20-040-20 HI22-20-040-40 HI22-20-040-50 HI22-20-040-60 HI22-20-100-10 HI22-20-100-20 HI22-20-100-40 HI22-20-100-50 HI22-20-100-60 HI22-25-010-15 HI22-25-010-30 HI22-25-010-50 HI22-25-010-60 HI22-25-025-10 HI22-25-025-105 HI22-25-025-15 HI22-25-025-30 HI22-25-025-50 HI22-25-025-60 HI22-25-050-10 HI22-25-050-105 HI22-25-050-15 HI22-25-050-30 HI22-25-050-50 HI22-25-050-60 HI22-25-100-10 HI22-25-100-105 HI22-25-100-15 HI22-25-100-30 HI22-25-100-50 HI22-25-100-60 HI22-32-010-10 HI22-32-010-105 HI22-32-010-15 HI22-32-010-30 HI22-32-010-50 HI22-32-010-60 HI22-32-025-10 HI22-32-025-105 HI22-32-025-15 HI22-32-025-30 HI22-32-025-50 HI22-32-025-60 HI22-32-050-10 HI22-32-050-105 HI22-32-050-15 HI22-32-050-30 HI22-32-050-50 HI22-32-050-60 HI22-32-100-10 HI22-32-100-105 HI22-32-100-15 HI22-32-100-30 HI22-32-100-50 HI22-32-100-60 HI22-32-150-10 HI22-32-150-105 HI22-32-150-15 HI22-32-150-30 HI22-32-150-50 HI22-32-150-60 HI22-53-010-10 HI22-53-010-15 HI22-53-010-30 HI22-53-025-15 HI22-53-025-30 HI22-53-025-50 HI22-53-025-60 HI22-53-050-10 HI22-53-050-15 HI22-53-050-30 HI22-53-050-50 HI22-53-050-60 HI22-53-100-10 HI22-53-100-15 HI22-53-100-30 HI22-53-100-50 HI22-53-100-60 HI22-53-100-75 HI22-53-150-10 HI22-53-150-15 HI22-53-150-30 HI22-53-150-50 HI22-53-150-60 HI22-53-150-75 HI22-05-005-2 HI22-05-005-2.5 HI22-05-005-5 HI22-10-010-10 HI22-10-010-15 HI22-10-010-5 HI22-10-020-10 HI22-10-020-15 HI22-10-020-5 HI22-10-040-10 HI22-10-040-15 HI22-10-040-5 HI22-15-015-10 HI22-15-015-15 HI22-15-015-20 HI22-15-015-30 HI22-15-015-5
Avantor® Hichrom HI-17 MS, GC Columns
Chromatography Columns
Really versatile general purpose polar phase for a wide range of applications. Ideal in confirmation analysis. HI-17 MS is an MS grade low bleeding cross-bonded column that ensures a long lifetime even under high temperature limits.
  • Phase composition: Crossbonded Silphenylene - Methyl Polysiloxane (17MS column selectivity) - MS grade
  • Long lifetime
Max. temperature limits may change depending on the film thickness.