Avantor® Hichrom, Column Frits
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HI-120EA 17.4 GBP
HI-120 HI-145
Avantor® Hichrom, Column Frits
Filters Precolumn Filters and Frits
Frit made of 316 grade stainless steel.
  • Helps in controlling porosity
A 2 μm porosity frit is recommended for retention of 5 to 20 μm packing material and 0,5 μm porosity frit is recommended for 3 to 4 μm packing materials.
Stainless steel with encapsulated PEEK used to prevent sample and eluent from entering dead volume space between the end fitting and also with the top of the column. This results in improved peak symmetry with 1,0 to 3,2 mm Int.Ø columns.