Acetonitrile for LC-MS

Supplier: Agilent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

G2453-85050
HEWLG2453-85050EA 0 GBP
HEWLG2453-85050
Acetonitrile for LC-MS
Acetonitrile

Agilent LC and LC/MS solvents and additives are manufactured to have the lowest levels of impurities and consistent lot-to-lot quality. Our solvents are designed specifically to work with Agilent instrumentation and applications.


Agilent solvents and additives are of the highest quality and are designed, developed, and manufactured to guarantee at least one year of shelf life.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.78
Flash Pt: 2
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
Merck Index: 13,00071

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR