Standard bare fused-silica capillaries
Supplier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
G1600-60211 G1600-60311 G1600-60411 G1600-61211 G1600-61311 G1600-61411 G1600-62211 G1600-62311 G1600-62411 G1600-63211 G1600-63311 G1600-63411 G1600-64211 G1600-64311 G1600-64411
HEWLG1600-60211EA 232 GBP
HEWLG1600-60211 HEWLG1600-60311 HEWLG1600-60411 HEWLG1600-61211 HEWLG1600-61311 HEWLG1600-61411 HEWLG1600-62211 HEWLG1600-62311 HEWLG1600-62411 HEWLG1600-63211 HEWLG1600-63311 HEWLG1600-63411 HEWLG1600-64211 HEWLG1600-64311 HEWLG1600-64411
Standard bare fused-silica capillaries
Electrophoresis Systems Capillary Electrophoresis Systems
Pre-aligned fused-silica capillaries from Agilent Technologies are designed and optimised for ease of use and reliability. All capillary ends are cut to a smooth, mirror-like finish. In addition, the polyimide outer coating is removed from the ends. These processes ensure minimal sample adsorption and help maintain sharp peak shapes. All capillaries have a pre-made detection 'window' and a built-in alignment stopper that allows rapid and precise insertion in the alignment interface.