Hollow cathode lamps for PerkinElmer atomic absorption spectrometers, single element
Supplier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8003-0712 8003-0713 8003-0714 8003-0715 8003-0716 8003-0888 8003-0889 8003-0890 8003-0891 8003-0892 8003-0893 8003-0894 8003-0897 8003-0898 8003-0899 8003-0900 8003-0901 8003-0904 8003-0905 8003-0906 8003-0907 8003-0908 8003-0909 8003-0910 8003-0911 8003-0912 8003-0913 8003-0914 8003-0915 8003-0916 8003-0917 8003-0918 8003-0919 8003-0920 8003-0922 8003-0923 8003-0924 8003-0925 8003-0926 8003-0927 8003-0928 8003-0929 8003-0930 8003-0931 8003-0932 8003-0934 8003-0936 8003-0937 8003-0938 8003-0939 8003-0941 8003-0942 8003-0943
HEWL8003-0712EA 1240 GBP
HEWL8003-0712 HEWL8003-0713 HEWL8003-0714 HEWL8003-0715 HEWL8003-0716 HEWL8003-0888 HEWL8003-0889 HEWL8003-0890 HEWL8003-0891 HEWL8003-0892 HEWL8003-0893 HEWL8003-0894 HEWL8003-0897 HEWL8003-0898 HEWL8003-0899 HEWL8003-0900 HEWL8003-0901 HEWL8003-0904 HEWL8003-0905 HEWL8003-0906 HEWL8003-0907 HEWL8003-0908 HEWL8003-0909 HEWL8003-0910 HEWL8003-0911 HEWL8003-0912 HEWL8003-0913 HEWL8003-0914 HEWL8003-0915 HEWL8003-0916 HEWL8003-0917 HEWL8003-0918 HEWL8003-0919 HEWL8003-0920 HEWL8003-0922 HEWL8003-0923 HEWL8003-0924 HEWL8003-0925 HEWL8003-0926 HEWL8003-0927 HEWL8003-0928 HEWL8003-0929 HEWL8003-0930 HEWL8003-0931 HEWL8003-0932 HEWL8003-0934 HEWL8003-0936 HEWL8003-0937 HEWL8003-0938 HEWL8003-0939 HEWL8003-0941 HEWL8003-0942 HEWL8003-0943
Hollow cathode lamps for PerkinElmer atomic absorption spectrometers, single element
Lights Hollow Cathode Lamps
Agilent hollow cathode lamps for PerkinElmer AA instruments are equivalent to the Lumina hollow cathode lamps and are compatible with PerkinElmer's entire range of atomic absorption spectrometers.
  • Designed for optimal performance and characterised by good sensitivity and spectral response
  • Lamps are element coded for automatic lamp recognition, preventing errors
  • Lamps for most elements use high quality quartz, except for Strontium (Sr) which uses borosilicate glass
All lamps are filled with neon gas for freedom from interfering lines at the mostMore Product Information