DV radial for PerkinElmer
Supplier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8003-0384 8003-0386 8003-0387 8003-0394 8003-0472
HEWL8003-0384EA 361 GBP
HEWL8003-0384 HEWL8003-0386 HEWL8003-0387 HEWL8003-0394 HEWL8003-0472
DV radial for PerkinElmer
Optical Windows
Agilent supplies are equivalent to PerkinElmer ICP-OES supplies for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Parts qualified on a genuine PerkinElmer system
Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime.