ICP-OES RF work/load coil for PerkinElmer
Supplier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8003-0379 8003-0380
HEWL8003-0379EA 1510 GBP
HEWL8003-0379 HEWL8003-0380
ICP-OES RF work/load coil for PerkinElmer
Chromatography Supplies Chromatography Spare Parts
These coils are equivalent to PerkinElmer ICP-OES coils for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the agilent stand behind warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage