Capillary column connectors, Press-fit
Supplier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5061-5804 5061-5805 5061-5806 5181-3398 500-1200 5190-6979
HEWL5061-5804EA 332 GBP
HEWL5061-5804 HEWL5061-5805 HEWL5061-5806 HEWL5181-3398 HEWL500-1200 HEWL5190-6979
Capillary column connectors, Press-fit
Fittings and Connectors
These Press-fit capillary connectors are laser milled to provide highly reproducible taper angles throughout the length, ensuring an excellent seal.