Split/splitless inlet seals
Supplier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
18740-20880 18740-60830 18740-60835 5182-9652 G1544-60585 5188-5367 5190-2209 5190-6144 5190-6145 5190-6149
HEWL18740-20880EA 81.6 GBP
HEWL18740-20880 HEWL18740-60830 HEWL18740-60835 HEWL5182-9652 HEWLG1544-60585 HEWL5188-5367 HEWL5190-2209 HEWL5190-6144 HEWL5190-6145 HEWL5190-6149
Split/splitless inlet seals
Seals Mechanical Seals
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
  • Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
  • High quality, leak-free seal
Delivery information: An inlet washer is provided with each inlet seal.
Order Now
SPECIFICATIONS
Environmentally Preferable