Forensic anion solutions kit
Supplier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5064-8208 5064-8209
HEWL5064-8208EA 1620 GBP
HEWL5064-8208 HEWL5064-8209
Forensic anion solutions kit
Standards Food Standards
This highly focused kit was developed specifically for the analysis of poisonous compounds, such as cyanide, azide, selenate, arsenate, and arsenite.
  • Requires minimal sample preparation
In cases of poisoning, analytical tools are needed to determine the identity of the toxins quickly and accurately. A rapid determination of anionic toxins in adulterated foods and beverages is possible using CE with indirect UV detection. Forensic and other anions can be detected within 15 minutes with minimal sample preparation.