Capillary columns, HP-1
Supplier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
19091-60312 19091S-004 19091Z-002 19091Z-005 19091Z-012 19091Z-015 19091Z-102 19091Z-105 19091Z-112 19091Z-115 19091Z-115E 19091Z-202 19091Z-205 19091Z-211 19091Z-212 19091Z-213 19091Z-213E 19091Z-215 19091Z-216 19091Z-216E 19091Z-231 19091Z-233 19091Z-233E 19091Z-236 19091Z-236E 19091Z-313 19091Z-331 19091Z-333 19091Z-411 19091Z-413 19091Z-413E 19091Z-416 19091Z-431 19091Z-433 19091Z-433E 19091Z-436 19091Z-513 19091Z-513E 19091Z-530 19091Z-530E 19091Z-613 19091Z-713 19091Z-713E 19091Z-716 19095S-100 19095S-100E 19095Z-020 19095Z-021 19095Z-023 19095Z-121 19095Z-121E 19095Z-123 19095Z-123E 19095Z-220 19095Z-221 19095Z-321 19095Z-323 19095Z-323E 19095Z-421 19095Z-421E 19095Z-423 19095Z-621 19095Z-623 19095Z-626 19095Z-221E 19095Z-627 19091Z-008 19091Z-102E 19091Z-112E 19091Z-411E 19095Z-321E 19095Z-423E 19095Z-623E 19091Z-577 19091S-004E 19091Z-105E 19091Z-215E 19091Z-431E 19091Z-716E 19095Z-021E 19095Z-023E 19095Z-623LTM 19091Z-313LTM 19091Z-577E 19091Z-713LTM 19095Z-021LTM 19095Z-023LTM 19095Z-121LTM 19095Z-123LTM
HEWL19091-60312EA 699 GBP
HEWL19091-60312 HEWL19091S-004 HEWL19091Z-002 HEWL19091Z-005 HEWL19091Z-012 HEWL19091Z-015 HEWL19091Z-102 HEWL19091Z-105 HEWL19091Z-112 HEWL19091Z-115 HEWL19091Z-115E 558-0404 HEWL19091Z-205 HEWL19091Z-211 HEWL19091Z-212 558-0395 HEWL19091Z-213E HEWL19091Z-215 HEWL19091Z-216 HEWL19091Z-216E HEWL19091Z-231 558-0396 HEWL19091Z-233E HEWL19091Z-236 HEWL19091Z-236E HEWL19091Z-313 HEWL19091Z-331 HEWL19091Z-333 HEWL19091Z-411 558-0397 HEWL19091Z-413E HEWL19091Z-416 HEWL19091Z-431 HEWL19091Z-433 HEWL19091Z-433E HEWL19091Z-436 HEWL19091Z-513 HEWL19091Z-513E HEWL19091Z-530 HEWL19091Z-530E HEWL19091Z-613 HEWL19091Z-713 HEWL19091Z-713E HEWL19091Z-716 HEWL19095S-100 HEWL19095S-100E HEWL19095Z-020 HEWL19095Z-021 HEWL19095Z-023 HEWL19095Z-121 HEWL19095Z-121E HEWL19095Z-123 HEWL19095Z-123E HEWL19095Z-220 HEWL19095Z-221 558-0402 HEWL19095Z-323 HEWL19095Z-323E HEWL19095Z-421 HEWL19095Z-421E HEWL19095Z-423 HEWL19095Z-621 558-0403 HEWL19095Z-626 HEWL19095Z-221E HEWL19095Z-627 HEWL19091Z-008 HEWL19091Z-102E HEWL19091Z-112E HEWL19091Z-411E HEWL19095Z-321E HEWL19095Z-423E HEWL19095Z-623E HEWL19091Z-577 HEWL19091S-004E HEWL19091Z-105E HEWL19091Z-215E HEWL19091Z-431E HEWL19091Z-716E HEWL19095Z-021E HEWL19095Z-023E HEWL19095Z-623LTM HEWL19091Z-313LTM HEWL19091Z-577E HEWL19091Z-713LTM HEWL19095Z-021LTM HEWL19095Z-023LTM HEWL19095Z-121LTM HEWL19095Z-123LTM
Capillary columns, HP-1
Chromatography Columns
Excellent general purpose column – "Industry Standard".
  • Non-polar
  • Wide range of applications
  • Superior performance for low molecular weight alcohols (
  • High temperature limit
  • Bonded and cross-linked
  • Solvent rinsable
100% Dimethylpolysiloxane
Similar phases: SPB-1, Rtx-1, BP-1, OV-1, OV-101, 007-1(MS), SP-2100, SE-30, ZB-1, AT-1, MDN-1, ZB-1
Order Now
SPECIFICATIONS
Environmentally Preferable