Heratherm® Stability Chamber
Supplier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ESS750
HERAESS750EA 18510 GBP
HERAESS750
Heratherm® Stability Chamber
Testing Chambers Humidity Chambers
Involved temperature and humidity controls, well suited for stability testing, shelf life testing, and environmental testing.
  • Broad relative humidity range to cover many applications, including humidity levels below ambient
  • Environmentally friendly water-efficient humidification system
  • Minimal condensate in standard applications
  • Integrated water tank, fixed water connection for in-house purified water option available
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard
  • Intuitive touchscreen interface with easy
More Product Information