Lids, Silicone Gaskets, and Screw Caps for Standard Cells
Supplier: HELLMA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01000110VE1072 01000150-VE572 011600-VE10734 011601-VE10734 011650-VE1072 011651-VE1072 010-001-00-10 010-001-1-10
HELL01000110VE1072EA 39.9 GBP
HELL01000110VE1072 HELL01000150-VE572 HELL011600-VE10734 HELL011601-VE10734 HELL011650-VE1072 HELL011651-VE1072 HELL010-001-00-10 HELL010-001-1-10
Lids, Silicone Gaskets, and Screw Caps for Standard Cells
Cuvettes Spectrophotometer Cuvettes
An assortment of lids, silicone gaskets, and screw caps designed for use with standard Hellma® cells.
  • Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers
  • Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism
  • Hellma® precision cells can be easily cleaned and reused and thus remain in use for a long time
Open screw caps are suitable for use with cell types 117.100, 117.100F, 117.104, 117.104F.
Closed screw caps are suitable for use with cell
More Product Information