PharmaFocus® Premium Reinforced Silicone Tubing
Supplier: Freudenberg Medical Europe GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
49464975 49464511 49464976 49456981 49446462 49464977 49446461 49464978 49450059 49464979 49464990
HELI49464975EA 692 GBP
HELI49464975 HELI49464511 HELI49464976 HELI49456981 HELI49446462 HELI49464977 HELI49446461 HELI49464978 HELI49450059 HELI49464979 HELI49464990
PharmaFocus® Premium Reinforced Silicone Tubing
Tubing Transfer Tubing
PharmaFocus® Premium Reinforced Silicone Tubing is available with single-braid reinforcement for enhanced performance in high pressure and vacuum applications. All reinforced silicone tubing is manufactured with an inner layer of platinum-cured silicone for ultra-pure fluid transfer and meets a diverse range of pharmaceutical and bioprocessing applications.
  • Performs in high purity fluid transfer and pump applications
  • Tube resilience and adaptability due to excellent flexibility
  • High visibility of fluid transport
  • Versatile processing with cut-to-length,
More Product Information