Culture media, ready to use, in tubes
Supplier: MERCK LIFE SCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1461410020 1464610010 1461810020 1461930100 1464800010 1461210020 1461390100 1461420020 1461750100 1461960100 1462080020 1462170100 1462190100 1462200100 1463270100 1464320100
HEIP1461410020EA 56.9 GBP
HEIP1461410020 HEIP1464610010 HEIP1461810020 HEIP1461930100 HEIP1464800010 HEIP1461210020 HEIP1461390100 HEIP1461420020 HEIP1461750100 HEIP1461960100 HEIP1462080020 HEIP1462170100 HEIP1462190100 HEIP1462200100 HEIP1463270100 HEIP1464320100
Culture media, ready to use, in tubes
Media for Microbiology Prepared Media for Microbiology
The current editions of the United States Pharmacopoeia (USP), European Pharmacopoeia (EP) and Japanese Pharmacopoeia (JP) include harmonized methods for the microbiological examination of non-sterile pharmaceutical products. These methods address the total aerobic count (TAMC and TYMC) as well as the test for specified micro-organisms. Harmonized methods facilitate and accelerate the release of pharmaceutical products.
  • A complete range of media required for testing all aerobic microorganisms in non-sterile pharmaceutical products
  • The composition of all media is in accordance with the requirements of the harmonized chapters of the Pharmacopoeia