Rotary Evaporators, Hei-VAP Expert Control
Supplier: Heidolph Instruments GmbH & Co.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
573-02100-03 573-02300-03 573-02305-03 573-02500-03 573-02600-03 573-02110-03 573-02310-03 573-02315-03 573-02510-03 573-02610-03 574-02100-03 574-02300-03 574-02305-03 574-02500-03 574-02600-03 574-02110-03 574-02310-03 574-02315-03 574-02510-03 574-02610-03 574-22600-00 574-23600-00 574-29300-00 574-37300-03 574-22600-03 574-23600-03 574-29300-03 574-32300-03 574-33300-03 574-38310-03
HEID573-02100-03EA 5580 GBP
HEID573-02100-03 HEID573-02300-03 HEID573-02305-03 HEID573-02500-03 HEID573-02600-03 HEID573-02110-03 HEID573-02310-03 HEID573-02315-03 HEID573-02510-03 HEID573-02610-03 HEID574-02100-03 HEID574-02300-03 531-1469 HEID574-02500-03 HEID574-02600-03 HEID574-02110-03 HEID574-02310-03 HEID574-02315-03 HEID574-02510-03 HEID574-02610-03 HEID574-22600-00 HEID574-23600-00 HEID574-29300-00 HEID574-37300-03 HEID574-22600-03 HEID574-23600-03 HEID574-29300-03 HEID574-32300-03 HEID574-33300-03 HEID574-38310-03
Rotary Evaporators, Hei-VAP Expert Control
Evaporators Rotary Evaporators
Hei-VAP Expert Control rotary evaporators allow central control of all process parameters: Vacuum, cooling temperature, rotation and heating bath temperature. The units have separate knobs for quick access: Right for direct vacuum control, the left one for rotation has a lock function to prevent accidental adjustment
  • 5" touch display for controlling the entire system including the vacuum and the cooling temperature
  • For precise monitoring, the pictogram on the display also illustrates active functions
  • Set values either via the touch display or the separate
More Product Information