Hei-TORQUE Overhead Stirrers, Gold 2 Package
Supplier: Heidolph Instruments GmbH & Co.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
501-62029-03
HEID501-62029-03EA 2710 GBP
HEID501-62029-03
Hei-TORQUE Overhead Stirrers, Gold 2 Package
Stirrers Overhead Stirrers
In order to provide the perfect end-to-end solution for powerful stirring and the simplest operation in the laboratory, we have added packages to the Hei-TORQUE product group. Each package contains a telescopic stand and a suitable clamp for easy use on lab tables.
Delivery information: Hei-TORQUE Ultimate 200 comes with telescope stand and clamp.