Pycnometer with thermometer, adjusted
Supplier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42572324 42572332 42572340 42572016 42572024 42572032 42572040
HECH42572324EA 239 GBP
HECH42572324 HECH42572332 HECH42572340 HECH42572016 HECH42572024 HECH42572032 HECH42572040
Pycnometer with thermometer, adjusted
Specific Gravity Bottles
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in capillary stopper
  • Very good chemical resistance
  • Very good temperature resistance
It includes pycnometer bottle and ground-in thermometer joint size 10/19 with blue filling.
Certifications: DIN ISO 3507