Clips for conical ground joints
Supplier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42386010 42386012 42386034 42386019 42386024
HECH42386010EA 60.1 GBP
HECH42386010 HECH42386012 HECH42386034 HECH42386019 HECH42386024
Clips for conical ground joints
Clamps NS Joint Clamps
Clips made of POM for conical ground joints.
  • Material does not scratch glass surfaces
  • Resistant to temperatures up to +150 °C