Measuring cylinders, low form, class B, soda-lime glass
Supplier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42205030 42205024 42205036 42205042 42206024 42205060 42205012 42205066 42205072 42205018 42205048 42206030
HECH42205030EA 16.3 GBP
HECH42205030 HECH42205024 HECH42205036 HECH42205042 HECH42206024 HECH42205060 HECH42205012 HECH42205066 HECH42205072 HECH42205018 HECH42205048 HECH42206030
Measuring cylinders, low form, class B, soda-lime glass
Cylinders Measuring Cylinders
Soda glass, low form, class B.
  • Calibrated to contain (TC, In)
  • Circular base or hexagonal plastic base , spout
  • Short line brown graduations
Certifications: DIN  EN ISO 4788