Gas washing bottles, Drechsel pattern, with head
Supplier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
41469012 41469018 41469024 41469030
HECH41469012EA 126 GBP
HECH41469012 HECH41469018 HECH41469024 HECH41469030
Gas washing bottles, Drechsel pattern, with head
Bottles Gas Washing Bottles
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Without filter disc.
Certifications: DIN 12596