Laboratory notebooks, waterproof
Supplier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HS8610B HS8610A
HEATHS8610BEA 38.9 GBP
HEATHS8610B HEATHS8610A
Laboratory notebooks, waterproof
Notebooks
For admissibility as court evidence, data on laboratory work, experiment details, and design and development. Use these notebooks to keep your indispensable data. Numbered pages include spaces for title, project number, book number, date, and witness signature.
  • The covers are water proof and chemical resistant
  • Contains periodic and conversion tables in the back