Cell counters
Supplier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HS5360A HS5360B
HEATHS5360AEA 462 GBP
HEATHS5360A HEATHS5360B
Cell counters
Cell Counters and Flow Cytometers Manual Cell Counters
These laboratory cell counters can measure cells such as basophils, eosinophils, myelocytes, juvenile, etc.
  • Each key records up to 999
  • Alarm at one hundred totals
  • Red and white buttons with grey housing
They can measure the number of cells such as Basophils (BASO), Eosinophils (EOSIN), Myelocytes (MYELO), Juvenile (JUVEN), Stab., Seg., Lymph., Mono., and total.