Counting chambers, Neubauer
Supplier: HAWKSLEY AND SONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AS2000
HAWKAS2000EA 368 GBP
HAWKAS2000
Counting chambers, Neubauer
Slides Counting Chambers
These cell counting chambers are moulded from a single piece of thermal and shock-resistant glass.
  • 2-cell
  • Optically polished platform
An H-shaped moat forms either one or two counting areas, or plateaus. Each plateau features an etched grid or ruling. Contact of the flat, polished cover glass surfaces with the cover glass supports produces an exact volume of fluid over the counting area.
The ruled surface is 0,1 mm below the cover glass, limiting the Volume of 1 Square mm of blood or fluid over a square mm at 0,1 cu.mm.
Certifications: BS 748
Order Now
SPECIFICATIONS
Environmentally Preferable