VWR® EML Digital Plus Test Sieve Shakers
Supplier: NEXOPART
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
510-0958EA 4520 GBP
510-0958 510-0955 510-0964 510-0961 510-0957 510-0954 510-0963 510-0960 510-1289 510-1298 510-1290 510-1296 510-1295 510-1293 510-1292 510-1299 HAVE209878547 HAVE209878493 HAVE209878516 HAVE209878561 HAVE209881981 HAVE209882001 HAVE209882025 HAVE209882049
VWR® EML Digital Plus Test Sieve Shakers
Shakers Sieve Shakers
These electromagnetically-driven test sieve shakers are designed for sieve analysis with optimum three-dimensional sieving action. The self-re-adjusting amplitude, which uses permanent acceleration measurement of the whole sieve tower, balances the oscillation of the shaker and its stand. This guarantees that amplitude stays the same, irrespective of the feed quantity and the installation position.
  • Memory for 10 test sieve parameter sets
  • Timer function: 0 to 99 minutes or continuous operation
  • Intermittent or continuous shaking operation
  • Digital display of
More Product Information