Technical pH buffer solutions in sachets, HI 50000 series
Supplier: Hanna
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HI50013-02 HI50012-01 HI50001-02 HI50005-02 HI50007-02 HI50011-02 HI50010-02 HI50004-02 HI50016-02
HANNHI50013-02EA 70.6 GBP
HANNHI50013-02 HANNHI50012-01 HANNHI50001-02 HANNHI50005-02 HANNHI50007-02 HANNHI50011-02 HANNHI50010-02 HANNHI50004-02 HANNHI50016-02
Technical pH buffer solutions in sachets, HI 50000 series
Buffers pH Calibration Buffers
These technical calibration buffers produced in accordance with ISO 3696/BS3978 standards using high purity salts, deionised water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers.
  • Technical buffer solutions with certificate
  • Accuracy of ±0,01 pH at 25 °C
  • Colour coded label for easy pH buffer value identification
Temperature chart of the actual pH value at various temperatures are printed on each sachets. Solutions are free from any dyes that may stain the reference cell of a pH electrode.
Certifications: Traceable to NIST Standard Reference Materials (SRMs).
Delivery information: These solutions are supplied with a certificate of analysis.