Combustion boats, Alsint 99,7
Supplier: MORGAN ADVANCED MATERIALS HALD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AL/115X16X9 ALSCH87X17X9 ALSCH115X16X9 ALSCH120X30X15 ALSCH160X40X21 ALSCH19X5X4 ALSCH200X15X15 ALSCH45X11X7 ALSCH52X7.5X6 ALSCH75X11X7
HALDAL/115X16X9EA 134 GBP
HALDAL/115X16X9 HALDALSCH87X17X9 459-2230 HALDALSCH120X30X15 HALDALSCH160X40X21 HALDALSCH19X5X4 HALDALSCH200X15X15 HALDALSCH45X11X7 HALDALSCH52X7.5X6 HALDALSCH75X11X7
Combustion boats, Alsint 99,7
Boats Combustion Boats
Alsint 99,7 (99,7 Al₂O₃).
  • High working temperature (<1700 °C), resistance to chemical attack, e.g. hydrogen and other reducing gases
  • High mechanical strength and highly impervious
  • High electrical resistivity