Precision™ General purpose water baths
Supplier: THERMO ELECTRON LED
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TSGP02 TSGP05 TSGP10 TSGP20 TSGP28 TSGP15D
HAKATSGP02EA 1060 GBP
HAKATSGP02 HAKATSGP05 HAKATSGP10 HAKATSGP20 HAKATSGP28 HAKATSGP15D
Precision™ General purpose water baths
Baths Water Baths, Unstirred
Precision™ general purpose water baths are designed to maintain water temperature from ambient to 100 °C.
  • Smaller footprint, compared to previous models, frees up valuable benchtop space
  • Advanced microprocessor controller is designed for extended functionality
  • Protects work with audible alarms
  • Conveniently save commonly used settings with four temperature presets
These rugged, high-performance baths range from 2 to 28 litres, including shallow models. Over-temperature safety circuitry is designed to prevent thermal runaway, while new auto-on and auto-More Product Information