Conductivity standards
Supplier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1440042 2307542 210542 2307442 1440049 2971849 1440026 LZW9700.99 LZW9710.99 LZW9720.99
HACH1440042EA 28.6 GBP
HACH1440042 HACH2307542 HACH210542 HACH2307442 HACH1440049 HACH2971849 HACH1440026 HACHLZW9700.99 HACHLZW9710.99 HACHLZW9720.99
Conductivity standards
Standards Conductivity Standards
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Order Now
SPECIFICATIONS
Environmentally Preferable