Ethylene glycol 0.05% in aqueous solution

Supplier: Hach
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

2183936
HACH2183936EA 19.8 GBP
HACH2183936
Ethylene glycol 0.05% in aqueous solution
Ethylene glycol
Formula: HOCH₂CH₂OH
Storage Temperature: Refrigerator
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR