Combined ecology test, AL-36B
Supplier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
180202
HACH180202EA 493 GBP
HACH180202
Combined ecology test, AL-36B
Environmental Test Kits
AL-36B can measure acidity, alkalinity, carbon dioxide (1,25 to 100 mg/L), dissolved oxygen (0,2 to 20 mg/L), hardness and pH (4 to 10).
  • Simple, accurate methods
  • Colour disc and drop-count titration methods
Designed to meet the portability needs of fish and wildlife conservation personnel.
Delivery information: Includes twelve reagents, clippers, bottles, sample cell with cap, colour disc, viewing tubes, comparator box, measuring tube, instructions and rugged carrying case.