Phenolphthalein 0,5% aqueous solution

Supplier: Hach
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

16232
HACH16232EA 22.1 GBP
HACH16232
Phenolphthalein 0,5% aqueous solution
Phenolphthalein
Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR