Borane-2-methylpyridine complex 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

H66642.06 H66642.14
H66642.06EA 48 GBP
H66642.06 H66642.14
Borane-2-methylpyridine complex 95%
Borane-2-methylpyridine complex

Purity: 95% CAS No.: 3999-38-0 Molecular Formula: C6H7N.BH3 Formula Weight: 106.96 Storage Temperature: 2°C to 8°C Flash Point: 100°(212°F) Physical Form: Solid Sensitivity: Moisture Sensitive Appearance: White Melting Point: 44-50° Solubility: Slightly sol. in water, cyclohexane, very sol. in methanol, THF, diglyme, and benzene UN No.: UN2813 MDL No.: MFCD07784361

Formula: C₆H₁₀BN
Melting Pt: 44…50 °C
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD07784361
CAS Number: 3999-38-0
ADR: 4.3,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR