2-Chloro-6-(trichloromethyl)pyridine 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H66418.03 H66418.06
H66418.03EA 32 GBP
H66418.03 H66418.06
2-Chloro-6-(trichloromethyl)pyridine 98%
2-Chloro-6-(trichloromethyl)pyridine

Purity: 98% CAS No.: 1929-82-4 Molecular Formula: C6H3Cl4N Formula Weight: 230.90 Flash Point: 100°(212°F) Physical Form: Powder Appearance: White Melting Point: 60-65° Beilstein Reference: 1618997 UN No.: UN3077 MDL No.: MFCD00072500

Formula: C₆H₃Cl₄N
Melting Pt: 60…65 °C
Flash Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00072500
CAS Number: 1929-82-4
EINECS: 217-682-2
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR