Fmoc-Asp(Ompe)-OH 98% (by HPLC)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H66281.06
H66281.06EA 200 GBP
H66281.06
Fmoc-Asp(Ompe)-OH 98% (by HPLC)
Fmoc-Asp(Ompe)-OH
Formula: C₂₅H₂₉NO₆
Storage Temperature: Freezer
MDL Number: MFCD10001333
CAS Number: 180675-08-5

Order Now

Specification Test Results

Assay (HPLC) 97.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR