6-Azaindole ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

H64458.03 H64458.06
H64458.03EA 48.1 GBP
H64458.03 H64458.06
6-Azaindole ≥97%
6-Azaindole

Purity: 98+% CAS No.: 271-29-4 Molecular Formula: C7H6N2 Formula Weight: 118.14 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Air and Light Sensitive Beilstein Reference: 109685 MDL No.: MFCD01686899

Formula: C₇H₆N₂
MW: 118.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01686899
CAS Number: 271-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR