2-chloro-5-iodo-3-(trifluoromethyl)pyridine 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine

H64329.03 H64329.06
H64329.03EA 48.6 GBP
H64329.03 H64329.06
2-chloro-5-iodo-3-(trifluoromethyl)pyridine 95%
2-chloro-5-iodo-3-(trifluoromethyl)pyridine

Purity: 95% CAS No.: 887707-25-7 Molecular Formula: C6H2ClF3IN Formula Weight: 307.44 Physical Form: Solid Sensitivity: Light Sensitive Melting Point: 43° Beilstein Reference: 10249610 MDL No.: MFCD12024302

Formula: C₆H₂ClF₃IN
MW: 307.439 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12024302
CAS Number: 887707-25-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR