5-Bromotryptamine 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3-(2-Aminoethyl)-5-bromoindole

H63653.MD H63653.03 H63653.06
H63653.MDEA 55.8 GBP
H63653.MD H63653.03 H63653.06
5-Bromotryptamine 97%
5-Bromotryptamine

Purity: 97% CAS No.: 3610-42-2 Molecular Formula: C10H11BrN2 Formula Weight: 239.11 Physical Form: Solid MDL No.: MFCD00130169

Formula: C₁₀H₁₁BrN₂
Storage Temperature: Ambient
MDL Number: MFCD00130169
CAS Number: 3610-42-2
EINECS: 222-779-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR